missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius MGC Grade Glass Microfiber Filters without Binder

Binder free MGC Grade Glass Microfiber Filters are suitable for analytical and gravimetric analyses. They are also used as prefilter.

$28.61 - $335.81

Specifications

Air Resistance 51 mbar
Color White
Format Disc
Material Borosilicate Microfiber Glass
Particle Retention 1.2 μm
View More Specs

Products 12
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
14-555-155
View Documents Promotion Details
Sartorius
FT31103185
18.5 cm
Pack of 50 Each for $207.64
 
14-555-156
View Documents Promotion Details
Sartorius
FT31103150
15 cm
Pack of 100 Each for $335.81
 
14-555-158
View Documents Promotion Details
Sartorius
FT31103110
11 cm
Pack of 100 Each for $198.71
 
14-555-157
View Documents Promotion Details
Sartorius
FT31103125
12.5 cm
Pack of 100 Each for $238.44
 
14-555-159
View Documents Promotion Details
Sartorius
FT31103100
10 cm
Pack of 100 Each for $168.90
 
14-555-160
View Documents Promotion Details
Sartorius
FT31103090
9 cm
Pack of 100 Each for $154.00
 
14-555-162
View Documents Promotion Details
Sartorius
FT31103055
5.5 cm
Pack of 100 Each for $87.53
 
14-555-161
View Documents Promotion Details
Sartorius
FT31103070
7 cm
Pack of 100 Each for $109.29
 
14-555-163
View Documents Promotion Details
Sartorius
FT31103050
5 cm
Pack of 100 Each for $81.86
 
14-555-164
View Documents Promotion Details
Sartorius
FT31103047
4.7 cm
Pack of 100 Each for $65.86
 
14-555-165
View Documents Promotion Details
Sartorius
FT31103025
2.5 cm
Pack of 100 Each for $51.76
 
14-555-166
View Documents Promotion Details
Sartorius
FT31103021
2.1 cm
Pack of 100 Each for $28.61
 
Description

Description

  • Applications include: Analysis of suspended solids in wastewater according to EN 872
Specifications

Specifications

51 mbar
Disc
1.2 μm
66 sec.
932°F to 1022°F
0.009 in.
White
Borosilicate Microfiber Glass
Non-sterile
MGC
500°C to 550°C
0.25 mm
SDS
missing translation for 'documents'

missing translation for 'documents'