missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius MGC Grade Glass Microfiber Filters without Binder

Binder free MGC Grade Glass Microfiber Filters are suitable for analytical and gravimetric analyses. They are also used as prefilter.

Supplier:  Sartorius FT31103100

 View more versions of this product

Catalog No. 14-555-159


Explore available promotions

Description

Description

  • Applications include: Analysis of suspended solids in wastewater according to EN 872
Specifications

Specifications

51 mbar
Glass Microfiber Filters without Binder
Borosilicate Glass Microfiber
Non-sterile
Ideal for critical analyses in which impurities can be washed out of the filter
932°F to 1022°F
0.009 in.
3.93 in.
White
Disc
1.2 μm
66 sec.
MGC
500°C to 550°C
0.25 mm
10 cm
SDS
Documents

Documents

Promotions

Promotions