missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius™ MGA Grade Glass Microfiber Filters w/o Binder

Manufacturer:  Sartorius™ FT31101185

 View more versions of this product

Catalog No. 14-555-180



Description

Description

Applications include: Clarification of buffer & reagent solutions, Filtration of algae cells.
Specifications

Specifications

36mbar
White
Clarification of buffer and reagent solutions
Filter Disc
1.6μm
0.009 in.
7.2 in.
Non-sterile
Borosilicate Microfiber Glass
18.5cm
40 sec.
MGA
500°C
0.25mm
932°F
SDS
Documents
Promotions

Promotions