Please login to your online account to display your discounted pricing

Sartorius™ Low Retention SafetySpace™ Filter Tips

Maximize sample recovery, pipetting precision and sample security. Sartorius™ Low Retention SafetySpace™ Filter Tips feature a liquid-repellent surface, additive-free filter and a perfect-sealing fit. Compatible with Sartorius Biohit™ pipettes. Sterile tips in a range of volumes are packed in single trays.

$190.38 - $210.47

Specifications

Product Type SafetySpace Filter Tips, Low Retention
For Use With (Equipment) Biohit™ pipettes and most other pipette brands
Autoclavable Yes
Filtered Yes
Sterilization Method E-beam
View More Specs
Products
Catalog Number Mfr. No. Color Code Length (English) Length (Metric) Volume (Metric) Price Quantity    

14-557-695

 
sartorius™
LHLF790101
Yellow Pack of 960 Each for $190.38

14-557-693

 
sartorius™
LHLF790011
Gray Pack of 960 Each for $190.38

14-557-698

 
sartorius™
LHLF791001
Blue Pack of 960 Each for $190.38

14-557-697

 
sartorius™
LHLF790301
Orange Pack of 960 Each for $190.38

14-557-694

 
sartorius™
LHLF790021
Yellow Pack of 960 Each for $190.38

14-557-696

 
sartorius™
LHLF790201
Green Pack of 960 Each for $190.38

14-557-699

 
sartorius™
LHLF791211
Purple Pack of 960 Each for $210.47
Description & Specifications

Specifications

Product Type SafetySpace Filter Tips, Low Retention
For Use With (Equipment) Biohit™ pipettes and most other pipette brands
Autoclavable Yes
Filtered Yes
Sterilization Method E-beam
Packaging Single Tray
Material Polypropylene tip, polyethylene filter

Low-retention tips for maximum sample recovery

  • Extremely even, liquid-repellent and hydrophobic surface optimizes the flow of liquids
  • Liquids slide easily off the chemically resistant surface, leaving very little residue behind
  • Ideal for fluids with low surface tension, such as viscous, detergent-containing liquids
  • Useful in PCR/real-time PCR, cloning, SDS-PAGE and other nucleic acid or protein analysis methods
Maximum safety and highest quality
  • Features a polyethylene filter without self-sealing additives that can interfere with the sample and results
  • Safety air gap between sample and filter reduces the risk of sample absorption into the filter
  • Filter protects against aerosol and liquid contamination and reduces pipette maintenance
  • E-beam pre-sterilized for sterile work
  • Certified DNase, RNase and endotoxin-free and compliant with ISO 9001 and ISO 13485
Ergonomically designed for precision and convenience
  • Perfect tip fitting and sealing to provide maximum precision and accuracy
  • Enables ergonomic and light tip attachment/ejection with Optiload feature of Sartorius pipettes
  • Compatible with Biohit™ Picus, eLINE™, mLINE™ and Proline™ Plus pipettes and most other brands
  • Color-coded trays and tips for easy matching with compatible Sartorius pipettes
  • Virgin polypropylene is 100% recyclable or can be incinerated as energy waste