missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius™ Glass Fiber (GF) Pre-filters, No Binder

Use in gravimetric analyses and as a depth filter for sterile filtration. Sartorius™ Glass Fiber (GF) Pre-filters, No Binder are made of borosilicate glass microfibers and combine fast flow rates with high load capacity. Ideal for relatively clean solutions, as it retains very fine particles.

Manufacturer:  Sartorius™ 1344047Q

Catalog No. 14-555-452



Description

Description

Binder-free glass microfiber filters are recommended for analytical and gravimetric analyses and also as prefilters. These filters combine fast flow rates with high load capacity and the retention of very fine particles.
  • A finer, binder-free filter disc for the prefiltration of relative clean solutions such as tissue culture media
    • It is placed directly on top of a membrane filter in a holder that specifies that disc diameter
    • Using a larger diameter disc will intrude under the sealing ring and cause leakage
    • Serial filtration may be necessary for difficult-to-filter liquids, such as serum
  • Biologically inert, resistant to most chemicals and temperature stable up to 500°C
  • Other filter characteristics:
    • Weight: 99.2g/m2
    • Filtration speed: 120mL/min.
    • Fulfills the requirements in EN 872:2005 (weigh loss)
Specifications

Specifications

Borosilicate Microfiber Glass
4.7cm
Prefiltration of clean solutions such as tissue culture media
Filter Disc
0.7μm
0.021 in.
Hydrophilic
932°F
White
Yes
4.1 to 4.4 sec. (Gurley air flow)
13440
500°C
550μm
1.8 in.
Non-sterile
Documents
Promotions

Promotions