Please login to your online account to display your discounted pricing
Sartorius™ Extraction Thimbles
Specifications
Bottom Shape Round
Sterile No
View More Specs
Products
Catalog Number Mfr. No. Grade Length (English) Max. Temperature (English) Material Price Quantity    
14-555-257
sartorius™
FT-1201-033094
30 3.7 in. 248°F Borosilicate Glass CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14-555-254
sartorius™
FT-1201-090200
30 7.8 in. 248°F Borosilicate Glass CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
Bottom Shape Round
Sterile No
  • Manufactured from cotton linters with a high alpha-cellulose content
  • High-purity thimbles that possess mechanical strength and outstanding retention capacity
  • Thermally stable up to 100°C
  • Inner diameter, outer length, round bottom and wall thickness defined according to the norm DIN 12449
  • Especially suited for all routine extractions: fat extractions, extraction of lipids from eggs and egg-containing products, softeners, additives, monomer components in plastics and rubber