Sartorius™ Entris™ Toploading Balances
When you need to get a heavy workload of repetitive applications done fast. Innocal Calibration Services avaliable on some models.
Manufacturer:  Sartorius™ ENTRIS22021SUS
 View more versions of this product
Catalog No. 14-557-456
Description
Description
  • White-backlit, high-contrast display with 15 mm digits; exceptionally easy to read under any room lighting conditions
  • New operating concept: easy configuration of the individual settings guided by short, menu-driven text prompts (choice of 5 languages) and simple navigation using cursor keys
  • RS-232C bidirectional interface port; optional Sartorius cable for connection of the balance to a USB port
  • Total ease of operation
  • All keys have positive click action
  • Clearly defined control panel
  • Monolithic weighing cell provides high thermal stability and extremely fast response times
  • Fast response time and display stability
  • Language-driven user interface
  • Unit toggle of up to 5 units
Applications
Percent weighing, unit toggle, animal weighing, net-total, calculation, density determination and counting
Specifications
Specifications
Toploading Balance
180mm
External Calibration
10mg
1.5 sec.
6.8 lb.
303 x 230 x 91mm (12.9 x 9.0 x 3.6 in.)
RS-232C, 25-pin
10 mg
30mg
ENTRIS2202-1S
3.1kg
180mm
2.2 kg