Sartorius™ Entris™ Precision Balances
Delivers effective and reliable weighing results in daily work. Sartorius™ Entris™ Precision Balances provide easy and clearly structured user interface, logical key assignment and excellent readability for error-free operation.
$1,005.42 - $2,555.63
Specifications
Products 8
Catalog Number Mfr. No. Readability Model Stabilization Time Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Readability Model Stabilization Time Capacity (Metric) Price Quantity  
14-557-449 View Documents Sartorius™
ENTRIS6231SUS
1 mg ENTRIS623-1S 1.0s 620g Each for $1,621.08
14-557-452 View Documents Sartorius™
ENTRIS1531SUS
1 mg ENTRIS153-1S 1.3s 150g Each for $1,005.42
14-557-450 Accessories View Documents Sartorius™
ENTRIS4231SUS
1 mg ENTRIS423-1S 1.0s 420g Each for $1,399.62
14-557-451 View Documents Sartorius™
ENTRIS3231S
1 mg ENTRIS323-1S 1.1s 320g Each for $1,178.16
14-559-020 View Documents Sartorius™
ENTRIS323I1SUS
1 mg ENTRIS323i-1S 1.1s 320g Each for $1,793.81
14-559-021 View Documents Sartorius™
ENTRIS423I1SUS
1 mg ENTRIS423i-1S 1.0s 420g Each for $2,152.58
14-559-019 View Documents Sartorius™
ENTRIS153I1SUS
1 mg ENTRIS153i-1S 1.3s 150g Each for $1,634.36
14-559-022 View Documents Sartorius™
ENTRIS623I1SUS
1 mg ENTRIS623i-1S 1.0s 620g Each for $2,555.63
Description
Description
  • White-backlit, high-contrast display with 15 mm digits; exceptionally easy to read under any room lighting conditions
  • New operating concept: easy configuration of the individual settings guided by short, menu-driven text prompts (choice of 5 languages) and simple navigation using cursor keys
  • RS-232C bidirectional interface port; optional Sartorius cable for connection of the balance to a USB port
  • Total ease of operation
  • All keys have positive click action
  • Clearly defined control panel
  • Monolithic weighing cell provides high thermal stability and extremely fast response times
  • Fast response time and display stability
  • Language-driven user interface
  • Unit toggle of up to 5 units
Applications
Percent weighing, unit toggle, animal weighing, net-total, calculation, density determination and counting
Specifications
Specifications
0.0
icon-pdf.png