Please login to your online account to display your discounted pricing

Sartorius™ Docu-pH+ Benchtop Meters

pH meter / mV meter

$887.32 - $887.32

Specifications

Accuracy (pH) ±0.005
Data Management Details Up to 500 data records
Detection Range 0 to 14pH
Display Type Color Graphic Display
Range (pH) 0 to 14
View More Specs
Products
Catalog Number Mfr. No. Accuracy (mV) Includes Price Quantity    

14-557-210

 
sartorius™
DOCUPHPLUS
±0.02 <1000mV Electrode retaining Arm, technical buffers, AC adapter, operating instructions Each for $887.32
Description & Specifications

Specifications

Accuracy (pH) ±0.005
Data Management Details Up to 500 data records
Detection Range 0 to 14pH
Display Type Color Graphic Display
Range (pH) 0 to 14
Resolution (pH) 0.001/0.01/0.1
For Use With (Application) pH
Probe Type pH
Range (mV) ±2000mV
Resolution (mV) 0.1/1mV
Temperature Compensation Automatic or Manual
Temperature Range (English) -41° to +221°F

DocuClip and Docu pH Meter let you close the reliability gap in electrochemistry. DocuClip and Docu pH Meter from Sartorius are setting new standards for documentation of results in electrochemistry.

  • Graphic display and soft keys for intuitive user guidance by plain-language prompts
  • Automatic temperature compensation, buffer recognition as well as recognition of various temperature sensors
  • Improved calibration routines support optimum procedural sequences in the laboratory
  • Calibration reminder
  • Memory for up to 500 data records
  • Data transfer over RS-232 to printer or PC
  • Automatic recognition of electrodes with DocuClip for unambiguous allocation of calibration data and complete documentation

DocuClip & DocuMeter let you close the reliability gap in electrochemistry
Laboratory work, particularly in the areas of chemical and pharmaceutical research, requires 100% traceable documentation. Conventional procedures, however, still hold some risks: Sensors that have been used may be mixed up. An electrode may have been used so often that it is no longer fully effective - but how can you tell? Manual recording of electrode identification is error-prone. Permanent allocation of identifying and calibration data to a particular pH electrode gives you complete reliability. It is no longer possible to mix up electrodes. The quality and effectiveness of electrodes can be checked at any time. Documentation of calibration data is fully automatic, making it both easy and reliable. It is entirely up to you whether you use your own electrodes or pre-calibrated electrodes from Sartorius. One click gives you complete reliability . . . with confidence supplied by Sartorius.