Please login to your online account to display your discounted pricing

Sartorius™ Docu-pH Benchtop Meters

pH meter / mV meter

$723.65 - $723.65

Specifications

Accuracy (mV) ±0.02 <1000mV
Accuracy (pH) ±0.005
Detection Range 0 to 14pH
Display Type Color Graphic Display
Range (pH) 0 to 14
View More Specs
Products
Catalog Number Mfr. No. Includes Temperature Compensation Price Quantity    

14-557-209

 
sartorius™
DOCUPH
Electrode retaining Arm, technical buffers, AC adapter, operating instructions Automatic Each for $723.65
Description & Specifications

Specifications

Accuracy (mV) ±0.02 <1000mV
Accuracy (pH) ±0.005
Detection Range 0 to 14pH
Display Type Color Graphic Display
Range (pH) 0 to 14
Resolution (pH) 0.001/0.01/0.1
For Use With (Application) pH
Probe Type pH
Range (mV) ±2000mV
Resolution (mV) 0.1/1mV
Temperature Range (English) -41° to +221°F

DocuClip™ and Docu pH Meter let you close the reliability gap in electrochemistry. DocuClip and Docu pH Meter from Sartorius are setting new standards for documentation of results in electrochemistry.

  • Graphic display and soft keys for intuitive user guidance by plain-language prompts
  • Automatic temperature compensation, buffer recognition as well as recognition of various temperature sensors
  • Improved calibration routines support optimum procedural sequences in the laboratory
  • Automatic recognition of electrodes with DocuClip for unambiguous allocation of calibration data

Docu-clip and Docu-pH/pH+ Meter let you close the traceability gap in electrochemistry
Laboratory work, particularly in the areas of chemical and pharmaceutical research, requires 100% traceable documentation. Conventional procedures, however, still hold some risks: Sensors that have been used may be mixed up. An electrode may have been used so often that it is no longer fully effective - but how can you tell? Manual recording of electrode identification is error-prone. Permanent allocation of identifying and calibration data to a particular pH electrode gives you complete reliability. It is no longer possible to mix up electrodes. The quality and effectiveness of electrodes can be checked at any time.