Sartorius™ DO Electrodes
Galvanic Dissolved Oxygen Probes
$528.01 - $528.01
Specifications
Probe Type Galvanic Dissolved Oxygen Probes
View More Specs
Products 1
Catalog Number Mfr. No. Sensor Types Price Quantity  
Catalog Number Mfr. No. Sensor Types Price Quantity  
14-557-899 View Documents Sartorius™
PYD022S
Integrated ATC NTC 30 K Ohm Each for $528.01
Description
Description
  • Membrane longevity up to 6 month
  • High signal resolution
  • Rapid measurement response
  • IP68
  • Includes calibration vessel
Specifications
Specifications
Galvanic Dissolved Oxygen Probes