Sartorius™ CultiBag RM Basic
Basic cultibags are designed for use in seed train and production applications without pH and DO control
$329.00 - $663.60
Specifications
Material EVA
For Use With For use in seed train and production applications without ph and do control
Includes Air inlet/outlet filter, sample port, fill/drain and spare ports, female and male luer, female male mpc connectors, ports with dip tubes
Tubing Material C-Flex™ 374 Silicon
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Volume (English) Working Capacity (English) Model Price Quantity  
Catalog Number Mfr. No. Volume (English) Working Capacity (English) Model Price Quantity  
14-558-209 Sartorius™
DBB001L
3.3 to 17 oz. 33.8 oz. CultiBag RM 1L Basic NA Each for $329.00
14-558-212 Sartorius™
DBB020L
67.6 to 338.1 oz. 5.2 gal. CultiBag RM 20L Basic NA Each for $564.20
14-558-210 Sartorius™
DBB002L
6.7 to 33.8 oz. 67.6 oz. CultiBag RM 2L Basic NA Each for $336.70
14-558-214 Sartorius™
DBB050L01US
1.32 to 6.6 oz. 13.2 gal. CultiBag RM 50L Basic GE Each for $627.20
14-558-213 Sartorius™
DBB050L
1.32 to 6.6 oz. 13.2 gal. CultiBag RM 50L Basic NA Each for $663.60
Specifications
Description
The CultiBag RM is a single use bioreactor bag available in multiple sizes and configurations depending on customer requirements. The inner layer of the disposable bioreactor bag is made of ethyl vinyl acetate (EVA), which shows an excellent biocompatibility.
Sensors
Precise and reliable single-use sensors for pH and DO come pre-installed and sterilized with the bag. The sensors are pre-calibrated and the calibration data is supplied together with the bag. After entering the data in the controller, the sensors are ready foruse. Calibration is fast and convenient. An optical fibre connects to the sensor patch, which is located at end of a sleeve in the bag. As there is no physical contact of the optical fiber to the inside of the bag, sterility is maintained at all times. The optical fibre transmits light of specific wavelengths to the sensor patch and returns the luminescence response from the sensor back to the measuring amplifier.
Flexibility and Configurations
The CultiBag RM is available in 1 L, 2 L, 10 L, 20 L, 50 L, 100 L, 200 L and 600 L bag sizes. Bags are available as basic, optical and perfusion pro configurations. Basic bags are designed for use in seed train and production applications without pH and DO control. Optical bags feature single use optochemical pH and DO sensor, which are already installed in everybag and calibrated. Used together with the BIOSTAT™ RM control tower, they allow high end cultivation with full process automation.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
EVA
For use in seed train and production applications without ph and do control
Air inlet/outlet filter, sample port, fill/drain and spare ports, female and male luer, female male mpc connectors, ports with dip tubes
C-Flex™ 374 Silicon