Sartorius™ Cubis™ MSU High-Capacity Toploader Balances
Speed and reliable results for the largest sample sizes
$7,927.54 - $9,473.94
Specifications
Electrical Requirements 100/240V, 50/60Hz
Connection Type RS-232 25 pin, RS232 9 pin, bluetooth, PS/2 port
Display Type LCD
Certifications/Compliance ISO, FDA/USP, GxP
View More Specs
Products 6
Catalog Number Mfr. No. Model Capacity Readability Stabilization Time Price Quantity  
Catalog Number Mfr. No. Model Capacity Readability Stabilization Time Price Quantity  
14-557-861 View Documents Sartorius™
MSU20201S000D0
20201S 20.2 kg 100 mg 1.5 sec. Each for $8,958.48
14-557-862 View Documents Sartorius™
MSU36201S000D0
36201S 36.2 kg 100 mg 1.5 sec. N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14-557-863 View Documents Sartorius™
MSU36201P000D0
36201P 10.2 to 36.2 kg 100 / 1000 mg 1.5 sec. Each for $9,473.94
14-557-864 View Documents Sartorius™
MSU70201S000D0
70201S 70.2 kg 100 mg 1.5 sec. N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14-557-865 View Documents Sartorius™
MSU36200S000D0
36200S 36.2 kg 1000 mg 1 sec. Each for $7,927.54
14-557-866 View Documents Sartorius™
MSU70200S000D0
70200S 70.2 kg 1000 mg 1 sec. N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
MSU High Capacity Toploader Balances are part of the Cubis modular equipment series, which incorporates a precisely-machined second generation monolithic weigh cell for increased accuracy and ultra-fast response for quick results with outstanding repeatability. It also allows customization to meet specific laboratory applications to fulfill the most stringent requirements in addition to the ability to choose the configuration to allow a user to invest in only what is necessary.
In the harsher environment of technical plants, the different container dimensions and larger material quantities demand that the weighing instruments be significantly more robust than those used in QC or research laboratories. In addition, higher demands are placed on their protection and cleaning. With an IP54 protection class top-quality and smooth surfaces, the Cubis high capacity balance is fully capable of withstanding these conditions. It consistently delivers speed and reliable results for the largest sample sizes, even under the most adverse conditions.
  • Offers the highest level of safety by providing uncompromising reliability of weighing results and conformity to standards
  • Short measurement times result in time gained—for every single measurement
  • Offers operator guidance when leveling is required—interactive prompts (in real-time) guide a user to quickly and properly position the spirit level by indicating which leveling foot to turn and in which direction)
  • Features isoCAL, a fully automatic calibration/adjustment function
  • Weighing pan is easily and quickly removable for cleaning
  • Q-Pan: Automatically minimizes off-center loading errors to maximize weighing accuracy
  • Offers a wide selection of accessories for extended options
  • Pan size: 400 x 300mm
MSU Display and Control Unit
  • Tiltable and removable large, high-resolution monochrome graphic display
  • Key operation; positive click action and precise activation of functions
  • Combines classic key-operated control with a wide range of performance features
  • Comes standard with built-in USB, 25-pin RS232C and Ethernet communication interfaces as well as integrated SD card reader for data storage and convenient data transfer
  • Q-Guide: Novel, wizard-supported user/task management for easy operation and programming
  • Ensure safety and reliability of results with Advanced Pharma Compliance via user/password management, audit trail function, alert messages and reminder functions
  • Applications include: Basic weighing, weighing in percent, averaging (animal weighing), mass unit conversation, counting, formulation, density determination, totalizing, checkweighing, second tare memory, individual identifiers (IDs), calculations, statistics, time controlled functions
Specifications
Specifications
100/240V, 50/60Hz
RS-232 25 pin, RS232 9 pin, bluetooth, PS/2 port
LCD
ISO, FDA/USP, GxP