Sartorius™ Cubis™ MSE Toploader Balances with Guide-Assisted Leveling
For users who need to weigh with the highest accuracy, but have lower needs in terms of complexity of a weighing process and data management.
$4,941.38 - $8,869.25
Specifications
Material Die-cast Metal, Stainless Steel
Electrical Requirements 100/240V 50/60Hz
Connection Type RS232 25 pin, RS232 9 pin, bluetooth, PS/2 port
Display Type LCD
Certifications/Compliance ISO, FDA/USP, GxP
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity  
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity  
14-557-377 Sartorius™
MSE8202S000D0
8202S ≤1 sec. 8200g 10mg Each for $7,103.98
14-557-376 Sartorius™
MSE10202S000D0
10202S ≤1 sec. 10,200g 10mg Each for $8,359.70
14-557-385 Sartorius™
MSE12201S000D0
12201S ≤1 sec. 12,200g 100mg Each for $5,498.33
14-557-386 Sartorius™
MSE8201S000D0
8201S ≤1 sec. 8200g 100mg Each for $4,941.38
14-557-374 Sartorius Corporation
MSE14202S000D0
14202S ≤1 sec. 14,200g 10mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14-557-375 Sartorius Corporation
MSE14202P000D0
14202P ≤1 sec. 3000 / 6200 / 14,200g 10 / 20 / 50mg Each for $8,869.25
14-557-025 Sartorius™
MSA3203S100DE
14202P ≤1 sec. 3500 / 7000 / 14,200g 10 / 20 / 50mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Description
MSE Toploader Balances are part of the Cubis modular equipment series, which incorporates a precisely-machined second generation monolithic weigh cell for increased accuracy and ultra-fast response for quick results with outstanding repeatability. Cubis is the first and only balance to automatically check, perform and document its exact leveling. It also allows customization to meet specific laboratory applications to fulfill the most stringent requirements in addition to the ability to choose the weighing module and draft shield to allow a user to invest in only what is necessary. 
  • Offers the highest level of safety by providing uncompromising reliability of weighing results and conformity to standards
  • Short measurement times result in time gained—for every single measurement
  • Comes standard with Q-Level, the automatic motorized leveling function—provides valuable support to always have a leveled balance (models >6.2kg offer operator guidance when leveling is required—interactive prompts (in real-time) guide a user to quickly and properly position the spirit level by indicating which leveling foot to turn and in which direction)
  • Features isoCAL, a fully automatic calibration/adjustment function
  • Q-Pan: Automatically minimizes off-center loading errors to maximize weighing accuracy
  • Interfaces, such as: Bluetooth™ (wireless), 9-pin RS232C with PS2 port (for the connection of a keyboard or barcode scanner) or an additional 25-pin RS232C port are optionally available
  • Without draft shield
  • Protected against dust and water
  • Offers a wide selection of accessories for extended options
  • Pan Size: 206 x 206mm (except for Model 5202S, which measures 140 x 140mm)
    MSE Display and Control Unit
  • Large, high-contrast LCD with clearly structured keys for precise activation of functions
  • Easy-to-understand menu guidance with short text prompts
  • Comes standard with built-in USB and 25-pin RS232C communication interfaces
  • Perfect for users who need to weigh with the highest accuracy, but who have less complex application/communication requirements
  • Applications include: Basic weighing, weighing in percent, averaging (animal weighing), mass unit conversation, counting, net/total formulation, density determination (buoyancy method only), totalizing, calculations
Description & Specifications
Specifications
Die-cast Metal, Stainless Steel
RS232 25 pin, RS232 9 pin, bluetooth, PS/2 port
ISO, FDA/USP, GxP
100/240V 50/60Hz
LCD