Sartorius™ Cubis™ MSE Precision Balances
The optimal solution for users who need to weigh with the highest accuracy, but demand lower complexity of weighing process and data management
$5,824.20 - $9,787.62
Specifications
Material Die-cast Metal, Stainless Steel
Electrical Requirements 100/240V 50/60Hz
Connection Type RS232 25 pin, RS232 9 pin, bluetooth, PS/2 port
Display Type LCD
Certifications/Compliance ISO, FDA/USP, GxP
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity  
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity  
14-557-365 Sartorius™
MSE5203S100DR
5203S ≤1 sec. 5200g 1mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14-557-358 Sartorius™
MSE3203S100DE
3203S ≤1 sec. 3200g 1mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14-557-359 Sartorius™
MSE2203S100DE
2203S ≤1 sec. 2200g 1mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14-557-364 Sartorius™
MSE323S100DE
323S ≤0.8 sec. 320g 1mg Each for $5,824.20
14-557-366 Sartorius™
MSE5203P100DR
5203P ≤1 sec. 1200 / 2400 / 5200g 1 / 2 / 5mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14-557-367 Sartorius™
MSE3203S100DR
3203S ≤1 sec. 3200g 1mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14-557-368 Sartorius™
MSE2203S100DR
2203S ≤1 sec. 2200g 1mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14-557-369 Sartorius™
MSE2203P100DR
2203P ≤1 sec. 1010 / 2200g 1 / 10mg Each for $9,787.62
14-557-370 Sartorius™
MSE1203S100DR
1203S ≤1 sec. 1200g 1mg Each for $8,863.33
14-557-371 Sartorius™
MSE623S100DR
623S ≤0.8 sec. 620g 1mg Each for $6,896.26
14-557-372 Sartorius™
MSE623P100DR
623P ≤0.8 sec. 150 / 300 / 620g 1 / 2 / 5mg Each for $5,960.48
14-557-373 Sartorius™
MSE323S100DR
323S ≤0.8 sec. 320g 1mg Each for $5,824.20
14-557-361 Sartorius Corporation
MSE1203S100DE
1203S ≤1 sec. 1200g 1mg Each for $8,863.33
36-102-3720 Sartorius™
MSE623S100DE
623S ≤0.8 sec. 620g 1mg Each for $6,807.74
14-557-356 Sartorius Corporation
MSE5203S100DE
5203S ≤1 sec. 5200g 1mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14-557-357 Sartorius Corporation
MSE5203P100DE
5203P ≤1 sec. 1200 / 2400 / 5200g 1 / 2 / 5mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14-557-360 Sartorius Corporation
MSE2203P100DE
2203P ≤1 sec. 1010 / 2200g 1 / 10mg Each for $9,787.62
14-557-363 Sartorius Corporation
MSE623P100DE
623P ≤0.8 sec. 150 / 300 / 620g 1 / 2 / 5mg Each for $6,049.00
Specifications
Description
MSE Precision Balances are part of the Cubis modular equipment series, which incorporates a precisely-machined second generation monolithic weigh cell for increased accuracy and ultra-fast response for quick results with outstanding repeatability. Cubis is the first and only balance to automatically check, perform and document its exact leveling. It also allows customization to meet specific laboratory applications to fulfill the most stringent requirements in addition to the ability to choose the weighing module and draft shield to allow a user to invest in only what is necessary.
  • Offers the highest level of safety by providing uncompromising reliability of weighing results and conformity to standards
  • Short measurement times result in time gained—for every single measurement
  • Comes standard with Q-Level, the automatic motorized leveling function—provides valuable support to always have a leveled balance
  • Features isoCAL, a fully automatic calibration/adjustment function
  • Q-Pan: Automatically minimizes off-center loading errors to maximize weighing accuracy
  • Interfaces, such as: Bluetooth™ (wireless), 9-pin RS232C with PS2 port (for the connection of a keyboard or barcode scanner) or an additional 25-pin RS232C port are optionally available
  • Draft shield and weigh pan can be quickly and easily disassembled and cleaned without compromising stability
  • Protected against dust and water
  • Offers a wide selection of accessories for extended options
  • Pan Size: 140 x 140mm
MSE Display and Control Unit
  • Large, high-contrast LCD with clearly structured keys for precise activation of functions
  • Easy-to-understand menu guidance with short text prompts
  • Comes standard with built-in USB and 25-pin RS232C communication interfaces
  • Perfect for users who need to weigh with the highest accuracy, but who have less complex application/communication requirements
  • Applications include: Basic weighing, weighing in percent, averaging (animal weighing), mass unit conversation, counting, net/total formulation, density determination (buoyancy method only), totalizing, calculations
† According to United States Pharmacopeia Chapter 41.
Description & Specifications
Specifications
Die-cast Metal, Stainless Steel
RS232 25 pin, RS232 9 pin, bluetooth, PS/2 port
ISO, FDA/USP, GxP
100/240V 50/60Hz
LCD