Sartorius™ Cubis™ Essential High-Capacity Balances
The right choice for users without complex operations who primarily want to perform ultra-precise weighing
$5,569.05 - $8,370.07
Specifications
Products ${productFamilyLength}
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity  
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity  
14559010 Sartorius™
MSE20201SED15
MSE20201S-ED15 ≤1.5 sec. 20 / 200g 100mg Each for $6,458.78
14559012 Sartorius™
MSE36200SED15
MSE36200S-ED15 ≤1 sec. 36 / 200g 1000mg Each for $5,569.05
14559011 Sartorius™
MSE36201SED15
MSE36201S-ED15 ≤1.5 sec. 36 / 200g 100mg Each for $8,370.07
Specifications
Description
Features
  • Large, high-contrast, liquid crystal Cubis MSE display
  • Easy-to-understand menu guidance with short text prompts
  • Clearly structured keys for precise activation of functions
  • Software support for fast leveling
  • Robust, monolithic weighing system technology for reliable measuring results
  • Easy calibration with isoCAL (internal automatic calibration / adjustment function)
  • Amply dimensioned, rectangular weighing pans
  • Data Interfaces: USB and RS232C; 3 optional ports are available as an accessory (Bluetooth, PS/2, RS232C)
  • Choose from 13 preconfigured premium Cubis Lab Balances, including semi-micro, analytical, precision and high-capacity balances
Applications
Unit conversion, isoCAL automatic calibration/adjustment function, density determination (buoyancy method only), calculations, averaging, net/total formulation, weighing in percent, counting, totalizing
Description & Specifications
Specifications
0.0