missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius Centrisart™ I Reverse Centrifugal Concentrators

Remove proteins from difficult to filter samples, even whole blood. Sartorius Centrisart I Reverse Centrifugal Concentrators performs centrifugal ultrafiltration against the direction of centrifugal force for fast and simple preparation of protein-free ultrafiltrates. PES and CTA membranes for 5,000 to 300,000 Dalton MWCO.

Manufacturer:  Sartorius 13209E

 View more versions of this product

Catalog No. 14-558-219



Description

Description

Ready-to-use unit for small volume centrifugal ultrafiltration
  • Separate proteins from low molecular weight substances in biological samples
  • Reverse centrifugal concentration process prevents premature blockage of the filter
  • Unique design can be used in fixed-angle or swinging-bucket rotors accepting standard 15 ml tubes
  • Ultrafiltration of 90% of the original sample volume is typically completed in about 30 minutes
  • Ultrafiltrate is collected in the floater insert tube, where it is accessible without removing the tube
Removal of proteins from biological samples is a critical step for many applications, including:
  • Drug binding studies
  • Determination of metabolites in serum
  • Protein removal from blood samples
  • Cleaning of liposomes
  • Virus removal
Specifications

Specifications

14 mm
93 mm
2,000xG (Fixed Angle Rotor); 2,500xG (Swinging Bucket Rotor)
Cellulose Triacetate, PES
Small volume centrifugal ultrafiltration of biological samples
Starter Pack: 3 of each 5000, 10,000, 20,000, 100,000 Da
12/Pk.
0.5 to 2.5 mL
SDS
Documents
Promotions

Promotions