Please login to your online account to display your discounted pricing

Sartorius™ BIOSTAT A™ Standard Equipment Packages, for Cell Culture Applications

Entry-level bioreactor/fermenter ideal for educational purposes

Specifications

For Use With (Application) Cell culture
For Use With (Equipment) Biostat A
Includes Control Tower w/motor, 4-gas aeration (air, O2, N2, CO2), 3 internal pumps, fixed speed, 2 external analog signal input
View More Specs
Products
Catalog Number Mfr. No. Capacity (English) Price Quantity    

14-558-125

 
sartorius™
CC1S
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

14-558-129

 
sartorius™
CC2S
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

14-558-133

 
sartorius™
CC5S
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

For Use With (Application) Cell culture
For Use With (Equipment) Biostat A
Includes Control Tower w/motor, 4-gas aeration (air, O2, N2, CO2), 3 internal pumps, fixed speed, 2 external analog signal input

Package Components

  • BIOSTAT A Control Tower (including motor)
  • Choice of UniVessel Glass 1, 2 or 5L, single-walled vessel
  • Laptop
  • Four-gas aeration (air, O2, N2, CO2)
  • Three internal pumps (fixed speed)
  • Digital probes for pH and DO
  • Foam/level probe
  • Two external analog signal inputs (4 to 20mA)
  • pH control (acid/CO2 base)
  • DO control
  • Combined foal/level control
  • Filter heater
  • Motor coupling
  • Three-blade segment impeller
  • Agitator shaft
  • Four-way addition port
  • Manual sampler
  • Universal Adapter
  • Harvest pipe (adjustable height)
  • Inoculation port
  • Air filter
  • Tool and accessory set