missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius™ BIOSTAT A™ Advanced Equipment Packages, for Cell Culture Applications

Entry-level bioreactor/fermenter ideal for educational purposes

Manufacturer:  Sartorius™ CC5A

 View more versions of this product

Catalog No. 14-558-135

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

 Package Components

  • BIOSTAT A Control Tower (including motor)
  • Choice of UniVessel Glass 1, 2 or 5L, single-walled vessel
  • Laptop
  • Four-gas aeration (air, O2, N2, CO2)
  • Three internal pumps (fixed speed)
  • Digital probes for pH and DO
  • Foam/level probe
  • Two external analog signal inputs (4 to 20mA)
  • pH control (acid/CO2 base)
  • DO control
  • Combined foal/level control
  • Filter heater
  • Motor coupling
  • Three-blade segment impeller
  • Agitator shaft
  • Heating blanket
  • Blind plugs for all ports
  • Micro-sparger
  • Four-way addition port
  • Manual sampler
  • Universal Adapter
  • Harvest pipe (adjustable height)
  • Inoculation port
  • Air filter
  • Tool and accessory set
Advanced Package also Provides:
  • Operation using tablet or smartphone (not included)
  • Substrate controller in the operator software for fed-batch process management
  • External, variable speed pump for substrate
Specifications

Specifications

5L
Biostat A
Bioreactor package
Cell culture
Advanced version for substrate controller/external feed pump; Operation via tablet and smartphone; 2 x spare analog inputs
SDS
Documents
Product Certifications