Sartorius Picus™ NxT Electronic Pipettes, 12 Channels
Designed to ease user's workload and to protect from repetitive strain injury (RSI)
Manufacturer:  Sartorius LH745461
 View more versions of this product
Catalog No. 14-557-833
Description
Description
Laboratory work requires repetition of similar pipetting steps that may cause strain on hand and arm, and thus may compromise results. To minimize this risk, a highly ergonomic electronic pipette should be used for minimum effort and strain. Sartorius Picus™ NxT are the most sophisticated and ergonomic electronic compact and light weight pipettes.
  • Password protection for stored programs
  • Very last liquid droplets dispensed by repeated blow-out for maximum recovery
  • Features one-touch operating-button and feather-light electronic tip ejection
  • Includes plate tracker for microwell plate dispensing
  • Electronic piston control and brake for precise pipetting
  • Safe-cone filters to prevent contamination
  • Adjustment wheel for volume setting and navigation
  • Range of pipetting modes for diluting and titrating to serial dispensing
  • Intuitive user interface in a choice of language options: English, French, German, Russian and Chinese
  • Certificate of accredited 3-point calibration
  • RFID-enabled to support asset management
  • Volume range: 0.2μL to 1200μL
Applications:
PCR and other DNA/RNA techniques, ELISA, Protein analysis and Cell culture
Certifications
CE Marked
Warranty
2 year
Specifications
Specifications
Orange
12
10 to 300 μL
SDS
Documents
Product Certifications
Accessories