missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius Optifit 50 to 1200μL Tips, Single Refill Pack

The perfect match for your pipetter

Manufacturer:  Sartorius 791202

 View more versions of this product

Catalog No. 14-559-488



Description

Description

Perfect tip fitting and sealing to the pipetter results in the best possible accuracy and precision. Compatible with Optiload feature in Picus, eLINE™, mLINE™ and Proline™ Plus pipetters, enabling ergonomic and light tip attachment and ejection.

  • Precision made tips
  • Provide best possible accuracy and precision
  • Compatible with most other pipetter makes
  • ISO 8 Clean Room manufacturing process
  • Made from highest quality virgin polypropylene
  • DNase, RNase and endotoxin-free certified
  • e-beam presterilized
  • Informative rack labeling improves tip identification and traceability: volume, product number, lot number
  • Racks, trays and tips are 100% recyclable
  • Air-tight plastic wrapping around the rack secures purity during transport and storage (plastic wrapping is regular waste)
  • 20 trays of 96 tips depending on tip volume
  • Tip trays are color-coded to indicate the matching, color-coded pipetter
  • Fully autoclavable at 121°C for 20 minutes

Certifications

CE certified. Manufactured according to ISO 9001 and ISO 13485.

Specifications

Specifications

Purple
Non-sterile
Autoclavable
2.81 in.
CE, ISO 9001, ISO 13485
Sartorius Picus™
Refill Pack
50 to 1200 μL
Purple
PP (Polypropylene)
7.15cm
DNase, RNase, ATP, and Endotoxin Free
Refill
SDS
Documents
Promotions

Promotions