missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius™ Biohit™ Optifit 50 to 1200μL Tips, Single Refill Pack

The perfect match for your pipetter

Manufacturer:  Sartorius™ 791203*

 View more versions of this product

Catalog No. 14-559-489



Description

Description

Perfect tip fitting and sealing to the pipetter results in the best possible accuracy and precision. Compatible with Optiload feature in Biohit Picus, eLINE™, mLINE™ and Proline™ Plus pipetters, enabling ergonomic and light tip attachment and ejection.

  • Precision made tips
  • Provide best possible accuracy and precision
  • Compatible with most other pipetter makes
  • ISO 8 Clean Room manufacturing process
  • Made from highest quality virgin polypropylene
  • DNase, RNase and endotoxin-free certified
  • e-beam presterilized
  • Informative rack labeling improves tip identification and traceability: volume, product number, lot number
  • Racks, trays and tips are 100% recyclable
  • Air-tight plastic wrapping around the rack secures purity during transport and storage (plastic wrapping is regular waste)
  • 20 trays of 96 tips depending on tip volume
  • Tip trays are color-coded to indicate the matching, color-coded Biohit pipetter
  • Fully autoclavable at 121°C for 20 minutes

Certifications

CE certified. Manufactured according to ISO 9001 and ISO 13485.

Specifications

Specifications

Purple
Sterile
Autoclavable
2.81 in.
CE, ISO 9001, ISO 13485
Sartorius™ Biohit™ Picus
Refill
50 to 1200 μL
Purple
PP (Polypropylene)
7.15cm
DNase, RNase, ATP, and Endotoxin Free
Sartorius™ Biohit™ eLINE™,Sartorius™ Biohit™ mLINE™,Sartorius™ Biohit™ Picus,Sartorius™ Biohit™ Proline™ Plus
Refill Pack
SDS
Documents
Promotions

Promotions