Please login to your online account to display your discounted pricing

Sartorius™ Biohit™ Optifit 10 to 1000uL Tips, Bulk in a Box

The perfect match for your pipetter

$23.41 - $23.41

Specifications

Certifications/Compliance DNase, RNase and endotoxin-free, CE, ISO 9001, ISO 13485
Autoclavable Yes
For Use With (Equipment) Optiload feature in Biohit™ Picus, eLINE™, mLINE™ and Proline™ Plus pipetters
Material Polypropylene
Volume (Metric) 10 to 1000μL
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

14-559-482

 
sartorius™
791005
Bulk in box Pack of 400 Each for $23.41
Case of 16 Pack for $374.56
Description & Specifications

Specifications

Certifications/Compliance DNase, RNase and endotoxin-free, CE, ISO 9001, ISO 13485
Autoclavable Yes
For Use With (Equipment) Optiload feature in Biohit™ Picus, eLINE™, mLINE™ and Proline™ Plus pipetters
Material Polypropylene
Volume (Metric) 10 to 1000μL
Color Blue

Perfect tip fitting and sealing to the pipetter results in the best possible accuracy and precision. Compatible with Optiload feature in Biohit Picus, eLINE™, mLINE™ and Proline™ Plus pipetters, enabling ergonomic and light tip attachment and ejection.

  • Precision made tips
  • Provide best possible accuracy and precision
  • Compatible with most other pipetter makes
  • ISO 8 Clean Room manufacturing process
  • Made from highest quality virgin polypropylene
  • Informative rack labeling improves tip identification and traceability: volume, product number, lot number
  • Racks, trays and tips are 100% recyclable
  • Packed in releasable bags in cardboard boxes
  • Fully autoclavable at 121°C for 20 minutes

CE certified. Manufactured according to ISO 9001 and ISO 13485.