Please login to your online account to display your discounted pricing
Sartorius™ Biohit™ Optifit 0.5 to 200μL Tips, Single Tray
The perfect match for your pipetter
$85.95 - $85.95
Specifications
Certifications/Compliance DNase, RNase and endotoxin-free, CE, ISO 9001, ISO 13485
Autoclavable Yes
For Use With (Equipment) Optiload feature in Biohit™ Picus, eLINE™, mLINE™ and Proline™ Plus pipetters
Material Polypropylene
Product Type Optifit Tip
View More Specs
Products
Catalog Number Mfr. No. Sterility Price Quantity    
14-559-467
sartorius™
790200
Nonsterile Pack of 960 Each for $85.95
Case of 8 Pack for $687.60
Description & Specifications
Specifications
Certifications/Compliance DNase, RNase and endotoxin-free, CE, ISO 9001, ISO 13485
Autoclavable Yes
For Use With (Equipment) Optiload feature in Biohit™ Picus, eLINE™, mLINE™ and Proline™ Plus pipetters
Material Polypropylene
Product Type Optifit Tip
Volume (Metric) 0.5 to 200μL
Perfect tip fitting and sealing to the pipetter results in the best possible accuracy and precision. Compatible with Optiload feature in Biohit Picus, eLINE™, mLINE™ and Proline™ Plus pipetters, enabling ergonomic and light tip attachment and ejection.
  • Precision made tips
  • Provide best possible accuracy and precision
  • Compatible with most other pipetter makes
  • ISO 8 Clean Room manufacturing process
  • Made from highest quality virgin polypropylene
  • DNase, RNase and endotoxin-free certified
  • Available as e-beam presterilized or nonsterilized
  • Informative rack labeling improves tip identification and traceability: volume, product number, lot number
  • Racks, trays and tips are 100% recyclable
  • Air-tight plastic wrapping around the rack secures purity during transport and storage (plastic wrapping is regular waste)
  • Tip trays are color-coded to indicate the matching, color-coded Biohit pipetter
  • Fully autoclavable at 121°C for 20 minutes
CE certified. Manufactured according to ISO 9001 and ISO 13485.