Sartorius™ Balance Wedge and Wedge Pro Data Transfer Software
$882.87 - $882.87
Specifications
For Use With (Application) Completely collects balance data and transfers it to any DOS/Windows application program
For Use With (Equipment) CPA balance, extended balance, Master Pro LA balance, SE and ME ultramicro and microbalance, M-power balance
Includes 5 ft. RS232 cable
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
14-557-515 Sartorius™
YSW06
Wedge Pro Each for $882.87
Specifications
Description
  • Includes 5 ft. cable
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Completely collects balance data and transfers it to any DOS/Windows application program
CPA balance, extended balance, Master Pro LA balance, SE and ME ultramicro and microbalance, M-power balance
5 ft. RS232 cable