Sartorius Balance Density Determination Kits
Use with Sartorius balances to determine the density of solids and liquids
Manufacturer:  Sartorius YDK01MS
 View more versions of this product
Catalog No. 14-557-545
Description
Description
All kits include
  • 2 Beakers (7mm and 55mm dia.)
  • Bar frame
  • Thermometer with retainer clip
  • Glass plummet
  • Sieve for immersing samples (pan hanger assembly)
  • Sample holder (pan hanger assembly)
  • Metal platform
CPA Kit includes
  • Gasket for ME models
  • Compensating disk for ME235S/P, ME254S models
  • 3 adapters
Cubis Kits include
  • Adapter for LA/LP models with analytical draft shield
Extended ED Kit includes
  • Adapters for LA/LP models: 1 with and 1 without analytical draft shield
  • Compensating disk for LA/LP2300D models
Specifications
Specifications
Cubis Balance; for Weighing Modules
Determining the density of solids and liquids
2 Beakers (7mm and 55mm dia.), bar frame, thermome