missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius™ Equipment and Accessories for Vacuum Filter Holders

Choose the best accessories to support your filtration applications. Sartorius™ Equipment and Accessories for Vacuum Filter Holders include an all-glass vacuum filter holder assembly plus a hose, hand-held pump with gauge, a 3-way valve, a 3-way tap and a stainless steel connector.

Manufacturer:  Sartorius™ 16623

 View more versions of this product

Catalog No. 14-555-549



Description

Description

Equipment and accessories for performing simple vacuum filtration
  • Glass vacuum filter holder assembly
    • Suitable for membrane filters with a diameter of 50 or 47mm
    • Includes a glass funnel, base and flask; metal clamp; and glass frit filter support
    • Vacuum-resistant flask with non-ground glass rim prevents contamination while pouring
  • Rubber vacuum hose
    • Thick walled rubber hose for connecting the system components
    • 100.6cm hose is useable for various sizes of hose nipples
  • 3-way valve
    • Used with a syringe to generate the vacuum necessary for filtration
    • Ideal when a permanently installed, powered vacuum generator is unavailable
  • Handheld vacuum pump with gauge
    • Hand-operated, PVC pump with a vacuum gauge and vacuum release lever
    • Includes 60cm of clear plastic tubing
    • Creates up to 80% vacuum inside and outside of the laboratory
  • 3-way tap
    • Stainless steel tap for use with multi-branch filtration system
    • Provides independent vacuum control for each funnel
  • Thread to Luer Lock Connector
    • Stainless steel connector metric
    • Features 12 x 1mm Male Thread To Male Luer Lock connections
Specifications

Specifications

Rubber
100.6cm length of thick walled rubber hose for connecting the system components
For vacuum filter holders and manifold systems
1 Pack
Hose nipples
Connecting system components, such as suction flasks, vacuum pumps, etc.
Rubber vacuum hose
SDS
Documents
Promotions

Promotions