Sartorius™ Accessories for the MD8 Airscan™ Air Sampler
Manufacturer: Sartorius™ 17016
Description
Accessories
Adapter, Connector - 14-555-869
- Connects airscan to isolator via flexible PVC hose and filter capsule
- DN 30 hose barb to 1-11 in. sanitary flange
- Connects airscan to isolator via silicone tubing and filter capsule
- DN 25 hose barb to 1-11 in. sanitary flange
- For culture media plates
- Replacement part for aluminum stack
- Intermediate (middle) part
- Stack of 10 open filter holders
- Portable flowrate measuring device for the MD8 series air samplers
- With PTFE membrane and sanitary flange inlet and outlet
- For sterile air filtration inserted between the MD8 airscan and isolator
- Stable case for the transport and the storage of unit and accessories
- For 1-11 in. sanitary flanges
- Aluminum
- Set of connectors for MD 8 (SM 16743/44)
- Aluminum
- Connects air sampler inlet to flexible hose
- Aluminum
- Connects flexible hose to filter holder/adapter
- Flexible hose to filter holder/adapter (S-F)
- For triclamp
- Holds 80mm gelatin filters
- Replacement part for the aluminum stack
- Adapter for gelatin filter units (gelatin disposables) to fit on the MD8 inlet
- 7.5 x 1.5mm
- For use with PC (dialog system software)
Specifications
For quantitative detection of airborne organisms, mainly in sterile areas of Class A/ B cleanrooms, isolators, or blow-fill-seal machines | |
Adapter | |
Airscan to isolator via silicone tubing |