missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius Accessories for Combisart™ Systems: Frits

Frits for use with Sartorius Combisart Systems

Manufacturer:  Sartorius 6980103

 View more versions of this product

Catalog No. 14-555-920



Description

Description

  • Stainless steel
  • For stainless steel manifolds
  • Choose from 47mm and 50mm

Specifications

Specifications

1.8 in.
1 Ea.
47mm
Stainless Steel manifold
Frit
4.7cm
1.85 in.
Ensure uniform distribution of microorganisms and particles on the membrane filter surface
Stainless Steel
SDS
Documents
Product Certifications
Promotions

Promotions