Please login to your online account to display your discounted pricing
Sartorius™ Absorbent Pads
presterilized in plastic magazines, which fit onto the Sartorius manual dispensing device
$73.00 - $187.00
Specifications
Product Type Absorbent Pad
View More Specs
Products
Catalog Number Mfr. No. Diameter (English) Outside Diameter (Metric) Outside Price Quantity    
14-555-730
sartorius™
1540050N
1.96 in. 5cm Pack of 100 Each for $73.00
14-555-732
sartorius™
1541047ALR
1.85 in. 4.7cm Pack of 1000 Each for $176.00
14-555-733
sartorius™
1541050ALR
1.96 in. Pack of 1000 Each for $187.00
Description & Specifications
Specifications
Product Type Absorbent Pad
Sartorius 1.4mm thick absorbent pads are wetted with the appropriate liquid culture medium before a membrane filter is placed on them. They come presterilized in plastic magazines, which fit onto the Sartorius manual dispensing device.
The absorbent pads are available in two diameters:
  • 47mm has approx. 3ml absorption capacity
  • 50mm with approx. 3.5ml absorption capacity