Sartorius Absorbent Pads
presterilized in plastic magazines, which fit onto the Sartorius manual dispensing device
Manufacturer:  Sartorius 1540050N
 View more versions of this product
Catalog No. 14-555-730
Description
Description
Sartorius 1.4mm thick absorbent pads are wetted with the appropriate liquid culture medium before a membrane filter is placed on them. They come presterilized in plastic magazines, which fit onto the Sartorius manual dispensing device.
The absorbent pads are available in two diameters:
  • 47mm has approx. 3ml absorption capacity
  • 50mm with approx. 3.5ml absorption capacity
Specifications
Specifications
1.96 in.
5cm
Absorbent Pad
Packaged in petridish