Sartorius™ Quantitative Grade 389 Filter Papers
Manufacturer:  Sartorius™ FT3102030
 View more versions of this product
Catalog No. 14-555-237
Description
Description
  • Ash-free (ash content ≤ 0.01 % according to DIN 540370)
  • Wet-strengthened
  • Color-coded box for easy selection
Specifications
Specifications
100 per pack
Disc
White
389
30mm
0.19mm