Sartorius Quantitative Grade 388 Filter Papers
These filter papers are used for quantitative and gravimetric analyses as well as for pressure or vacuum filtration. They are made out of 100 % cotton linters with an α-cellulose content of > 98 % and are acid-washed to make the papers ashless and achieve high purity.
Supplier:  Sartorius FT3101090
 View more versions of this product
Catalog No. 14-555-244
Explore available promotions
Description
Description
Quantitative Grade 388 Filter Papers
  • Ash-free (ash content ≤ 0.01 % according to DIN 540370)
  • Wet-strengthened
  • Color-coded box for easy selection
Specifications
Specifications
Black
10 to 15 μm
388
0.21 mm
Disc
10 sec.
100/Pk.
90 mm