Sartorius™ Quantitative Grade 388 Filter Papers
Manufacturer:  Sartorius™ FT3101110
 View more versions of this product
Catalog No. 14-555-243
Description
Description
  • Ash-free (ash content ≤ 0.01 % according to DIN 540370)
  • Wet-strengthened
  • Color-coded box for easy selection
Specifications
Specifications
100 per pack
Disc
Black
388
110mm
0.21mm