missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius™ Qualitative Grade 293 Filter Papers

$12.00 - $40.00

Specifications

Format Disc
Grade 293
Quantity 100 per pack
Thickness 0.15mm
View More Specs

Products 7
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
14-555-033
View Documents
Sartorius™
FT3211185
185mm
Pack of 100 Each for $40.00
 
14-555-034
View Documents
Sartorius™
FT3211150
150mm
Pack of 100 Each for $26.00
 
14-555-035
View Documents
Sartorius™
FT3211125
125mm
Pack of 100 Each for $21.00
 
14-555-037
View Documents
Sartorius™
FT3211090
90mm
Pack of 100 Each for $12.00
 
14-555-036
View Documents
Sartorius™
FT3211110
110mm
Pack of 100 Each for $15.00
 
14-555-038
View Documents
Sartorius™
FT3211070
70mm
Pack of 100 Each for $12.00
 
14-555-039
View Documents
Sartorius™
FT3211055
55mm
Pack of 100 Each for $12.00
 
Description

Description

Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.
Specifications

Specifications

Disc
293
100 per pack
0.15mm
SDS