Sartorius Qualitative Grade 292 a Filter Papers
Manufacturer:  Sartorius FT3215150
 View more versions of this product
Catalog No. 14-555-027
Description
Description
  • Pure cotton linters or cotton linters with refined pulp
  • No additives, such as wet-strengthening agents
Specifications
Specifications
150 mm
292 a
100/Pk.
Disc
4 to 7μm
0.19 mm