Sartorius Qualitative Grade 1292 Filter Papers
Manufacturer:  Sartorius FT3210240
 View more versions of this product
Catalog No. 14-555-040
Description
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.
Specifications
Specifications
240 mm
1292
100/Pk.
Disc
5 to 8μm
0.17 mm