missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius™ Qualitative Grade 1292 Filter Papers

$12.00 - $64.00

Specifications

Format Disc
Grade 1292
Quantity 100 per pack
Thickness 0.17mm
View More Specs

Products 8
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
14-555-040
View Documents
Sartorius™
FT3210240
240mm
Pack of 100 Each for $64.00
 
14-555-041
View Documents
Sartorius™
FT3210185
185mm
Pack of 100 Each for $40.00
 
14-555-044
View Documents
Sartorius™
FT3210110
110mm
Pack of 100 Each for $15.00
 
14-555-042
View Documents
Sartorius™
FT3210150
150mm
Pack of 100 Each for $26.00
 
14-555-046
View Documents
Sartorius™
FT3210070
70mm
Pack of 100 Each for $12.00
 
14-555-045
View Documents
Sartorius™
FT3210090
90mm
Pack of 100 Each for $12.00
 
14-555-043
View Documents
Sartorius™
FT3210125
125mm
Pack of 100 Each for $21.00
 
14-555-047
View Documents
Sartorius™
FT3210055
55mm
Pack of 100 Each for $12.00
 
Description

Description

Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.
Specifications

Specifications

Disc
1292
100 per pack
0.17mm
SDS