Sartorius™ Qualitative Grade 1291 Filter Papers
$10.00 - $53.00
Specifications
Grade 1291
Particle Retention 2 to3μm
Quantity 100 per pack
Thickness 0.16mm
View More Specs
Products 8
Catalog Number Mfr. No. Diameter (Metric) Format Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Format Price Quantity  
14-555-048 View Documents Sartorius™
FT3209240
240mm Disc Pack of 100 Each for $53.00
14-555-049 View Documents Sartorius™
FT3209185
185mm Disc Pack of 100 Each for $33.00
14-555-050 View Documents Sartorius™
FT3209150
150mm Disc Pack of 100 Each for $22.00
14-555-052 View Documents Sartorius™
FT3209110
110mm Disc Pack of 100 Each for $14.00
14-555-051 View Documents Sartorius™
FT3209125
125mm Disc Pack of 100 Each for $18.00
14-555-053 View Documents Sartorius™
FT3209090
90mm Disc Pack of 100 Each for $11.00
14-555-054 View Documents Sartorius™
FT3209070
70mm Disc Pack of 100 Each for $10.00
14-555-055 View Documents Sartorius™
FT3209055
55mm Disc Pack of 100 Each for $10.00
Description
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.
Specifications
Specifications
1291
2 to3μm
100 per pack
0.16mm