Sartorius Qualitative Grade 1291 Filter Papers
Manufacturer:  Sartorius FT3209070
 View more versions of this product
Catalog No. 14-555-054
Description
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.
Specifications
Specifications
70 mm
1291
100/Pk.
Disc
2 to 3μm
0.16 mm