Learn More
Sartorius Qualitative Grade 1290 Filter Papers
These qualitative filter papers are essentially used for analytical purposes and routine analyses, whenever no gravimetric analyses are required. They are wet-strengthened and can be used for pressure and vacuum filtration.
Supplier: Sartorius FT3208125
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.Specifications
| Disc | |
| Cotton Linters, Refined Pulp | |
| 3 to 5 μm | |
| 1290 | |
| 0.21 mm | |
| 125 mm |
| Conical Funnel | |
| 3 to 5 μm | |
| 100 sec. | |
| 100/Pk. | |
| Qualitative Filter Paper |