Sartorius™ Sartolab™ P20 Plus Filter Systems
Filter tissue culture solutions quickly, economically and reliably. Sartorius™ Sartolab™ P20 Plus Filter Systems are sterile, ready-to-use units. A large filtration area and automatic de-aeration ensures high flow rates. Available with PES or SFCA membrane, different connectors or with a prefilter to process up to 10L.
Manufacturer:  Sartorius™ 18068D
 View more versions of this product
Catalog No. 14-555-990
Description
Description
Filter tissue culture solutions quickly, economically and reliably. Sartorius™ Sartolab™ P20 Plus Filter Systems are sterile, ready-to-use units. A large filtration area and automatic de-aeration ensures high flow rates. Available with PES or SFCA membrane, different connectors or with a prefilter to process up to 10L.
The new way for reliable sterile filtration
  • Ready-to-use unit with a large filtration area (20cm2) for fast filtration
  • Quickly and reliably sterilizes media and aqueous solutions with optimal throughputs
  • Automatically vents any trapped air through the PTFE membrane-protected vent ports
  • Extraordinarily favorable price-performance ratio for a cost-effective solution
  • The total filterable volume can often be doubled due to an integrated binder-free, glass fiber pre-filter
  • High-quality construction; every batch is rigorously tested to ensure sterility and integrity
Specifications
Specifications
Polyethersulfone
Glass pre-filter, Filling bell
Luer Lock / Hose nipple
Reliable sterile filtration of tissue culture solutions
5 bar
0.2μm
10 Pack
0.9mL
Sterile
USP, Class VI
20cm2
3 bar
PES
Sartolab P20 Plus Filter System
100mL to 10L