Sartorius Sartolab™ P20 Plus Filter Systems
Filter tissue culture solutions quickly, economically and reliably. Sartorius Sartolab™ P20 Plus Filter Systems are sterile, ready-to-use units. A large filtration area and automatic de-aeration ensures high flow rates. Available with PES or SFCA membrane, different connectors or with a prefilter to process up to 10L.
Manufacturer:  Sartorius 18056D
 View more versions of this product
Catalog No. 14-555-988
Description
Description
Filter tissue culture solutions quickly, economically and reliably. Sartorius Sartolab™ P20 Plus Filter Systems are sterile, ready-to-use units. A large filtration area and automatic de-aeration ensures high flow rates. Available with PES or SFCA membrane, different connectors or with a prefilter to process up to 10L.
The new way for reliable sterile filtration
  • Ready-to-use unit with a large filtration area (20cm2) for fast filtration
  • Quickly and reliably sterilizes media and aqueous solutions with optimal throughputs
  • Automatically vents any trapped air through the PTFE membrane-protected vent ports
  • Extraordinarily favorable price-performance ratio for a cost-effective solution
  • The total filterable volume can often be doubled due to an integrated binder-free, glass fiber pre-filter
  • High-quality construction; every batch is rigorously tested to ensure sterility and integrity
Specifications
Specifications
USP, Class VI
20 cm2
3 bar
10 Pack
0.9 mL
100 mL to 10 L
0.2 μm
100 mL to 10 L
Hose nipple / Hose nipple
Reliable sterile filtration of tissue culture solutions
5 bar
No pre-filter, Filling bell
100 mL to 10 L
Surfactant-free Cellulose Acetate (SFCA)
Sterile