missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tocris™ Sarpogrelate hydrochloride

Selective 5-HT2A antagonist

$218.74 - $915.10

Specifications

Product Type Sarpogrelate hydrochloride
Inhibitors Sarpogrelate hydrochloride
Molecular Weight 465.97
Purity >99
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
373910R Tocris™
RDS373910
10mg Each for $218.74
373950R Tocris™
RDS373950
50mg Each for $915.10
Specifications

Description

Selective 5-HT2A receptor antagonist (pKi values are 8.52, 7.43 and 6.57 for 5-HT2A, 5-HT2C and 5-HT2B receptors respectively). Displays selectivity over 5-HT1, 5-HT3, 5-HT4 H1, H2, M3, α1-adrenergic, α2-adrenergic and β-adrenergic receptors. Displays cardioprotective activity in vivo.
  • Chemical Name: Succinic acid mono-(1-dimethylaminomethyl-2-(2-[2-(3-methoxyphenyl)ethyl]phenoxy)ethyl) ester hydrochloride
  • Formula: C24H31NO6.HCl
  • CAS Number: 135159-51-2
  • Solubility: Soluble to 25mM in water and to 100mM in DMSO
  • Purity: >99
Description & Specifications

Specifications

Sarpogrelate hydrochloride
Sarpogrelate hydrochloride
465.97
>99