missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sarcosine methyl ester hydrochloride, +97%, ACROS Organics™

$435.61 - $435.61

Chemical Identifiers

CAS 13515-93-0
Assay Percent Range 97%
Molecular Formula C4H9NO2·HCl
Formula Weight 139.58
MDL Number MFCD00038876
Chemical Name or Material Sarcosine methyl ester, hydrochloride
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC397502500 SDS Thermo Fisher Scientific
AC397502500
Glass bottle 250g Each for $435.61
AC397500500 SDS Thermo Fisher Scientific
AC397500500
Glass bottle 50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

13515-93-0
C4H9NO2·HCl
MFCD00038876
≥96.0%
97%
139.58
Sarcosine methyl ester, hydrochloride

Specifications

13515-93-0
97%
Authentic
MFCD00038876
White to beige
250g
≥96.0%
Crystals or Crystalline Powder
C4H9NO2·HCl
139.58
Glass bottle
110° to 120°C
Sarcosine methyl ester, hydrochloride

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.