Please login to your online account to display your discounted pricing

Alfa Aesar™ Sarcosine hydrochloride, 99%

$52.34 - $170.86

Chemical Identifiers

Chemical Name or Material Sarcosine hydrochloride
CAS 637-96-7
Molecular Formula CH3NHCH2CO2H·HCl
Formula Weight 125.56
MDL Number MFCD00012609
Merck Index 14,8373
Synonym Methylaminoacetic acid hydrochloride; N-Methylglycine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2553614

 
alfa aesar™
B2553614
25g Each for $52.34

AAB2553622

 
alfa aesar™
B2553622
100g Each for $170.86
Description & Specifications

Specifications

Formula Weight 125.56
Chemical Name or Material Sarcosine hydrochloride
TSCA TSCA
Assay 99%
Molecular Formula CH3NHCH2CO2H·HCl
MDL Number MFCD00012609
CAS 637-96-7
Merck Index 14,8373
Synonym Methylaminoacetic acid hydrochloride; N-Methylglycine hydrochloride
Melting Point ≈173°C (dec.)
Quantity 25g

Applications
Sarcosine hydrochloride is used in organic synthesis.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.