Please login to your online account to display your discounted pricing

Alfa Aesar™ Sarcosine anhydride, 99%

$29.24 - $88.05

Chemical Identifiers

Chemical Name or Material Sarcosine anhydride
CAS 5076-82-4
Molecular Formula C6H10N2O2
Formula Weight 142.16
MDL Number MFCD00006519
Beilstein 123513
Synonym 1,4-Dimethylpiperazine-2,5-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1019103

 
alfa aesar™
A1019103
1g Each for $29.24

AAA1019106

 
alfa aesar™
A1019106
5g Each for $88.05
Description & Specifications

Specifications

Formula Weight 142.16
Chemical Name or Material Sarcosine anhydride
Assay 99%
Molecular Formula C6H10N2O2
MDL Number MFCD00006519
CAS 5076-82-4
Beilstein 123513
Synonym 1,4-Dimethylpiperazine-2,5-dione
Melting Point 142° to 147°C
Quantity 1g