missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sarcosine, 98%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 157740050

 View more versions of this product

Catalog No. AC157740050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Sarcosine
(25% aq. soln.) clear, colorless to slightly yellow
≥97.5% (total base)
C3H7NO2
Authentic
Glass Bottle
2% max.
5g
107-97-1
Crystals or Powder
98%
CH3NHCH2CO2H
89.09
N-Methylglycine
Colorless or white to yellow

Chemical Identifiers

98%
CH3NHCH2CO2H
N-Methylglycine
≥97.5% (total base)
C3H7NO2
89.09
Sarcosine