Please login to your online account to display your discounted pricing

Alfa Aesar™ Sarcosinamide hydrochloride, 97%

Chemical Identifiers

Chemical Name or Material Sarcosinamide hydrochloride
CAS 5325-64-4
Linear Formula CH3NHCH2CONH2·HCl
Formula Weight 124.57
MDL Number MFCD00058283
Beilstein 3678388
Synonym H-Sar-NH2.HCl; 2-(Methylamino)-acetamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6599303

 
alfa aesar™
H6599303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6599306

 
alfa aesar™
H6599306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH65993MD

 
alfa aesar™
H65993MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 124.57
MDL Number MFCD00058283
Chemical Name or Material Sarcosinamide hydrochloride
Physical Form Solid
Assay 97%
Linear Formula CH3NHCH2CONH2·HCl
CAS 5325-64-4
Storage Note 1 -30° to -10°C
Beilstein 3678388
Synonym H-Sar-NH2.HCl; 2-(Methylamino)-acetamide hydrochloride
Color Off white to pale yellow
Quantity 1g
Safety and Handling

-30° to -10°C