Please login to your online account to display your discounted pricing

Saquinavir mesylate, Tocris™

HIV protease inhibitor

$102.98 - $436.69

Chemical Identifiers

Chemical Name or Material Saquinavir mesylate
Assay Percent Range >99%
Molecular Formula C38H50N6O5·CH4O3S
Formula Weight 766.95
Synonym (2S)-N1-[(1S,2R)-3-[(3S,4aS,8aS)-3-[[(1,1-dimethylethyl)amino]carbonyl]octahydro-2(1H)-isoquinolinyl]-2-hydroxy-1-(phenylmethyl)propyl]-2-[(2-quinolinylcarbonyl)amino]butanediamide methanesulfonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

441810

 
r & d systems
RDS441810
10mg Each for $102.98

441850

 
r & d systems
RDS441850
50mg Each for $436.69
Description & Specifications

Specifications

Formula Weight 766.95
Quantity 10mg
Chemical Name or Material Saquinavir mesylate
Assay Percent Range >99%
Molecular Formula C38H50N6O5·CH4O3S
CAS 149845-06-7
Synonym (2S)-N1-[(1S,2R)-3-[(3S,4aS,8aS)-3-[[(1,1-dimethylethyl)amino]carbonyl]octahydro-2(1H)-isoquinolinyl]-2-hydroxy-1-(phenylmethyl)propyl]-2-[(2-quinolinylcarbonyl)amino]butanediamide methanesulfonate
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C